Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
102128691
Gene Name
interleukin 26
Gene Aliases
-
Chromosome Location
Chr.11: 67239841 - 67267625 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571479.2 XP_005571536.1 3 - 4 604 143

View Full Product Details