Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT18
Entrez Gene ID
102138807
Gene Name
nudix hydrolase 18
Gene Aliases
-
Chromosome Location
Chr.8: 22243502 - 22252163 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005563258.2 XP_005563315.1 4 - 5 1181 61

View Full Product Details