Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPK3B
Entrez Gene ID
102127227
Gene Name
uroplakin 3B
Gene Aliases
-
Chromosome Location
Chr.3: 46923556 - 46930224 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549319.2 XP_005549376.1 2 - 3 564 137

View Full Product Details