Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21A
Entrez Gene ID
102141044
Gene Name
methyltransferase like 21A
Gene Aliases
-
Chromosome Location
Chr.12: 97184933 - 97244097 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574084.2 XP_005574141.1 3 - 4 618 104
XM_005574085.2 XP_005574142.1 3 - 4 408 104
XM_015432663.1 XP_015288149.1 4 - 5 367 104
XM_015432664.1 XP_015288150.1 3 - 4 453 104
GenBank mRNA AB063050.1 - - 251 104

View Full Product Details