Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMIE
Entrez Gene ID
102145868
Gene Name
transmembrane inner ear
Gene Aliases
-
Chromosome Location
Chr.2: 101722814 - 101731899 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005546917.2 XP_005546974.1 2 - 3 312 123

View Full Product Details