Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM83C
Entrez Gene ID
102138594
Gene Name
family with sequence similarity 83 member C
Gene Aliases
-
Chromosome Location
Chr.10: 66754765 - 66761101 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005568822.2 XP_005568879.1 3 - 4 909 136

View Full Product Details