Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTN4RL2
Entrez Gene ID
102139905
Gene Name
reticulon 4 receptor like 2
Gene Aliases
-
Chromosome Location
Chr.14: 16144822 - 16162010 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015434707.1 XP_015290193.1 2 - 3 584 84

View Full Product Details