Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT26
Entrez Gene ID
102140971
Gene Name
keratin 26
Gene Aliases
-
Chromosome Location
Chr.16: 52419637 - 52425714 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584114.1 XP_005584171.1 4 - 5 980 80

View Full Product Details