Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINGO4
Entrez Gene ID
102119649
Gene Name
leucine rich repeat and Ig domain containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 100160440 - 100165811 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541860.2 XP_005541917.1 1 - 2 402 81

View Full Product Details