Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD34A
Entrez Gene ID
102140420
Gene Name
ankyrin repeat domain 34A
Gene Aliases
-
Chromosome Location
Chr.1: 104362566 - 104367725 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015430004.1 XP_015285490.1 2 - 3 1882 82

View Full Product Details