Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL4D
Entrez Gene ID
102124666
Gene Name
ADP ribosylation factor like GTPase 4D
Gene Aliases
-
Chromosome Location
Chr.16: 55075671 - 55077628 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005584355.2 XP_005584412.1 1 - 2 138 100

View Full Product Details