Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNS3
Entrez Gene ID
102142113
Gene Name
potassium voltage-gated channel modifier subfamily S member 3
Gene Aliases
-
Chromosome Location
Chr.13: 93052368 - 93118675 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005576491.2 XP_005576548.1 2 - 3 336 67
XM_005576492.2 XP_005576549.1 2 - 3 283 67
XM_005576493.2 XP_005576550.1 2 - 3 343 67

View Full Product Details