Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNB1
Entrez Gene ID
102123118
Gene Name
potassium voltage-gated channel subfamily B member 1
Gene Aliases
-
Chromosome Location
Chr.10: 81150971 - 81264730 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005569322.2 XP_005569379.1 1 - 2 729 143

View Full Product Details