Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
PNMA2
Entrez Gene ID
102124250
Gene Name
PNMA family member 2
Gene Aliases
-
Chromosome Location
Chr.8: 26933901 - 26940907 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348450.1 NP_001335379.1 1 - 1 33 81
XM_015454370.1 XP_015309856.1 1 - 1 69 81
GenBank mRNA CJ443242.1 - 1 - 1 60 81

View Full Product Details