Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARC
Entrez Gene ID
102130416
Gene Name
activity regulated cytoskeleton associated protein
Gene Aliases
-
Chromosome Location
Chr.8: 144128018 - 144131519 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564182.2 XP_005564239.1 2 - 3 2218 77

View Full Product Details