Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102125097
Entrez Gene ID
102125097
Gene Name
60S ribosomal protein L36a
Gene Aliases
RPL36A
Chromosome Location
Chr.X: 98543152 - 98548243 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005596080.2 XP_005596137.2 1 - 2 229 142

View Full Product Details