Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC6
Entrez Gene ID
102130456
Gene Name
sialic acid binding Ig like lectin 6
Gene Aliases
-
Chromosome Location
Chr.19: 52268423 - 52285770 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005590124.2 XP_005590181.1 3 - 4 2207 153

View Full Product Details