Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102140954
Entrez Gene ID
102140954
Gene Name
interferon alpha-8
Gene Aliases
IFNA8
Chromosome Location
Chr.15: 57347414 - 57348640 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581619.2 XP_005581676.1 1 - 1 354 105

View Full Product Details