Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMX1
Entrez Gene ID
102128184
Gene Name
empty spiracles homeobox 1
Gene Aliases
-
Chromosome Location
Chr.13: 36925017 - 36945514 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575611.2 XP_005575668.1 2 - 3 1051 164

View Full Product Details