Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ORAI1
Entrez Gene ID
102129315
Gene Name
ORAI calcium release-activated calcium modulator 1
Gene Aliases
-
Chromosome Location
Chr.11: 125646437 - 125661871 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005572459.2 XP_005572516.1 1 - 1 136 67

View Full Product Details