Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTL5
Entrez Gene ID
102127629
Gene Name
retrotransposon Gag like 5
Gene Aliases
RGAG4
Chromosome Location
Chr.X: 69172213 - 69177111 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593935.2 XP_005593992.1 1 - 1 2074 108
XM_005593936.2 XP_005593993.1 1 - 1 2074 108
GenBank mRNA AB060816.1 - 1 - 1 1284 108

View Full Product Details