Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXB1
Entrez Gene ID
102130018
Gene Name
homeobox B1
Gene Aliases
-
Chromosome Location
Chr.16: 33882131 - 33885201 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583571.2 XP_005583628.1 1 - 2 1265 69

View Full Product Details