Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPTN
Entrez Gene ID
102141349
Gene Name
repetin
Gene Aliases
-
Chromosome Location
Chr.1: 99759967 - 99764974 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015457126.1 XP_015312612.1 1 - 2 154 68

View Full Product Details