Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP62
Entrez Gene ID
102125639
Gene Name
ZFP62 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.6: 180859938 - 180877567 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558821.2 XP_005558878.1 1 - 2 105 87

View Full Product Details