Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNPY1
Entrez Gene ID
102117343
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
-
Chromosome Location
Chr.3: 188414927 - 188451520 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015448337.1 XP_015303823.1 2 - 3 350 171
XM_015448338.1 XP_015303824.1 2 - 3 772 171

View Full Product Details