Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC48A1
Entrez Gene ID
102128684
Gene Name
solute carrier family 48 member 1
Gene Aliases
-
Chromosome Location
Chr.11: 46505907 - 46529184 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570666.2 XP_005570723.1 3 - 4 387 69
XM_005570667.2 XP_005570724.1 3 - 4 443 69
XM_005570668.2 XP_005570725.1 3 - 4 449 69
XM_015430836.1 XP_015286322.1 2 - 3 210 69

View Full Product Details