Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL7B
Entrez Gene ID
102131630
Gene Name
methyltransferase like 7B
Gene Aliases
-
Chromosome Location
Chr.11: 54575842 - 54578913 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005571128.1 XP_005571185.1 1 - 2 760 85

View Full Product Details