Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXD12
Entrez Gene ID
102128170
Gene Name
homeobox D12
Gene Aliases
-
Chromosome Location
Chr.12: 65061977 - 65074476 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573552.2 XP_005573609.1 1 - 2 1690 83

View Full Product Details