Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSMP
Entrez Gene ID
102138312
Gene Name
microseminoprotein, prostate associated
Gene Aliases
-
Chromosome Location
Chr.15: 43356031 - 43357516 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005581351.2 XP_005581408.1 1 - 2 476 91

View Full Product Details