Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACOT6
Entrez Gene ID
102145211
Gene Name
acyl-CoA thioesterase 6
Gene Aliases
-
Chromosome Location
Chr.7: 138245049 - 138252434 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561723.2 XP_005561780.1 3 - 3 1157 103

View Full Product Details