Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAS4
Entrez Gene ID
102133639
Gene Name
breast carcinoma amplified sequence 4
Gene Aliases
-
Chromosome Location
Chr.10: 82624737 - 82714766 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005569346.2 XP_005569403.1 5 - 5 706 120
XM_005569347.2 XP_005569404.1 4 - 4 571 120

View Full Product Details