Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPN1LW
Entrez Gene ID
102137107
Gene Name
opsin 1 (cone pigments), long-wave-sensitive
Gene Aliases
-
Chromosome Location
Chr.X: 151153753 - 151168160 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283209.1 NP_001270138.1 4 - 5 748 93
GenBank mRNA AF158968.1 - 4 - 5 748 93
AF158975.1 - 4 - 5 748 93

View Full Product Details