The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283461.1 | NP_001270390.1 | 1 - 2 | 821 | 66 |
XM_015452267.1 | XP_015307753.1 | 1 - 2 | 860 | 66 | |
XM_015452270.1 | XP_015307756.1 | 2 - 3 | 506 | 66 | |
XM_015452273.1 | XP_015307759.1 | 2 - 3 | 447 | 66 | |
GenBank mRNA | AB046635.1 | - | 1 - 2 | 406 | 66 |
AB062973.1 | - | 1 - 2 | 821 | 66 |