Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSRA
Entrez Gene ID
102142280
Gene Name
methionine sulfoxide reductase A
Gene Aliases
-
Chromosome Location
Chr.8: 10226737 - 10653119 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283603.1 NP_001270532.1 4 - 5 581 102
XM_005562595.2 XP_005562652.1 4 - 5 1559 102
XM_005562600.2 XP_005562657.1 3 - 4 1434 102
XM_005562601.2 XP_005562658.1 4 - 5 2622 102
XM_005562602.2 XP_005562659.1 5 - 6 2688 102
XM_015454378.1 XP_015309864.1 4 - 5 1812 102
XM_015454379.1 XP_015309865.1 3 - 4 1752 102
XM_015454382.1 XP_015309868.1 6 - 7 3852 102
XM_015454383.1 XP_015309869.1 3 - 4 1709 102
XM_015454384.1 XP_015309870.1 4 - 5 1584 102
XM_015454385.1 XP_015309871.1 4 - 5 3641 102
XM_015454386.1 XP_015309872.1 4 - 5 1577 102
XM_015454387.1 XP_015309873.1 5 - 6 3714 102
XM_015454388.1 XP_015309874.1 3 - 4 972 102
XM_015454389.1 XP_015309875.1 3 - 4 2850 102
XM_015454390.1 XP_015309876.1 4 - 5 1561 102
XM_015454391.1 XP_015309877.1 4 - 5 1558 102
XM_015454392.1 XP_015309878.1 4 - 5 1559 102
GenBank mRNA AB220441.1 - 4 - 5 581 102

View Full Product Details