Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOB1
Entrez Gene ID
101866049
Gene Name
NIN1 (RPN12) binding protein 1 homolog
Gene Aliases
-
Chromosome Location
Chr.20: 58053781 - 58067272 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283542.1 NP_001270471.1 4 - 5 419 68
GenBank mRNA AB169707.1 - 4 - 5 419 68

View Full Product Details