Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP50
Entrez Gene ID
101865081
Gene Name
ubiquitin specific peptidase 50
Gene Aliases
-
Chromosome Location
Chr.7: 27478799 - 27530595 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283999.1 NP_001270928.1 5 - 6 984 85
XM_005559539.2 XP_005559596.1 5 - 6 988 85
XM_015452403.1 XP_015307889.1 5 - 6 990 85
XM_015452404.1 XP_015307890.1 5 - 6 984 85
XM_015452405.1 XP_015307891.1 5 - 6 973 85
XM_015452406.1 XP_015307892.1 5 - 6 867 85
XM_015452407.1 XP_015307893.1 4 - 5 795 85
GenBank mRNA AB169252.1 - 5 - 6 984 85

View Full Product Details