Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKA2
Entrez Gene ID
101865188
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
FAM33A
Chromosome Location
Chr.16: 44659770 - 44702140 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285190.1 NP_001272119.1 3 - 4 307 106
GenBank mRNA AB168504.1 - 3 - 4 307 106

View Full Product Details