Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCD
Entrez Gene ID
101925980
Gene Name
IQ motif containing D
Gene Aliases
-
Chromosome Location
Chr.11: 116916239 - 116944311 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283765.1 NP_001270694.1 4 - 5 1302 91
XM_015430670.1 XP_015286156.1 3 - 4 1366 91
GenBank mRNA AB168312.1 - 4 - 5 1302 91

View Full Product Details