Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUPT4H1
Entrez Gene ID
101925811
Gene Name
SPT4 homolog, DSIF elongation factor subunit
Gene Aliases
-
Chromosome Location
Chr.16: 43865327 - 43872749 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284797.1 NP_001271726.1 4 - 5 1503 74
GenBank mRNA AB168256.1 - 4 - 5 1503 74

View Full Product Details