Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD9
Entrez Gene ID
101926579
Gene Name
PDZ domain containing 9
Gene Aliases
-
Chromosome Location
Chr.20: 20813340 - 20830809 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284799.1 NP_001271728.1 2 - 3 261 110
GenBank mRNA AB168207.1 - 2 - 3 261 110

View Full Product Details