Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRK1
Entrez Gene ID
102120479
Gene Name
killer cell lectin-like receptor subfamily K, member 1
Gene Aliases
nkg2D
Chromosome Location
Chr.11: 10683039 - 10699940 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283284.1 NP_001270213.1 4 - 5 295 98
XM_015430167.1 XP_015285653.1 5 - 6 537 98
GenBank mRNA AJ426429.1 - 4 - 5 295 98

View Full Product Details