Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNCG
Entrez Gene ID
102121834
Gene Name
synuclein gamma
Gene Aliases
-
Chromosome Location
Chr.9: 48761654 - 48769283 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283870.1 NP_001270799.1 4 - 5 452 67
XM_015455894.1 XP_015311380.1 6 - 7 825 67
GenBank mRNA AB220471.1 - 4 - 5 452 67

View Full Product Details