Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEATR9
Entrez Gene ID
101926787
Gene Name
HEAT repeat containing 9
Gene Aliases
-
Chromosome Location
Chr.16: 32225765 - 32241597 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284066.1 NP_001270995.1 4 - 5 570 87
XM_005583485.2 XP_005583542.1 4 - 5 607 87
GenBank mRNA AB168985.1 - 4 - 5 570 87

View Full Product Details