Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C9
Entrez Gene ID
102115450
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CYP2C43
Chromosome Location
Chr.9: 93472905 - 93526478 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287625.1 NP_001274554.1 4 - 5 672 91
XM_005566002.2 XP_005566059.1 4 - 5 737 91
XM_005566006.2 XP_005566063.1 4 - 5 613 91
GenBank mRNA DQ074806.1 - 4 - 5 672 91

View Full Product Details