Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HBB
Entrez Gene ID
101926697
Gene Name
hemoglobin, beta
Gene Aliases
-
Chromosome Location
Chr.14: 65216098 - 65217690 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283367.1 NP_001270296.1 2 - 3 372 103
GenBank mRNA AB171600.1 - 2 - 3 372 103

View Full Product Details