Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102138191
Entrez Gene ID
102138191
Gene Name
cortistatin
Gene Aliases
CORT
Chromosome Location
Chr.1: 217727589 - 217747248 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015442744.1 XP_015298230.1 3 - 4 483 105

View Full Product Details