Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGAS
Entrez Gene ID
102133276
Gene Name
cyclic GMP-AMP synthase
Gene Aliases
MB21D1
Chromosome Location
Chr.4: 100469214 - 100498370 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015449182.1 XP_015304668.1 4 - 5 1435 115

View Full Product Details