Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102130062
Entrez Gene ID
102130062
Gene Name
WAP four-disulfide core domain protein 6
Gene Aliases
WFDC6
Chromosome Location
Chr.10: 77250720 - 77256255 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015457700.1 XP_015313186.1 1 - 2 250 110

View Full Product Details