Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRS1
Entrez Gene ID
102117978
Gene Name
insulin receptor substrate 1
Gene Aliases
-
Chromosome Location
Chr.12: 116526101 - 116589153 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574453.2 XP_005574510.1 1 - 2 4004 73

View Full Product Details