Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102141672
Entrez Gene ID
102141672
Gene Name
WD repeat-containing protein 87
Gene Aliases
WDR87
Chromosome Location
Chr.19: 38277130 - 38302727 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589071.2 XP_005589128.2 4 - 5 3311 67
XM_015441296.1 XP_015296782.1 4 - 5 3357 67

View Full Product Details